Tryptic Soy Agar (TSA) Blood Agar Base, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C5220
C5221
C5222
C5223
CA89405-200EA
166.12
CAD
CA89405-200
CA89405-202
CA89405-204
CA89405-206
Tryptic Soy Agar (TSA) Blood Agar Base, CRITERION™, Hardy Diagnostics
Media for Microbiology
Dehydrated Media for Microbiology
CRITERION™ TSA Blood Agar Base is used with blood for isolation of fastidious microorganisms when hemolytic reactions are important
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
This medium is also recommended for use in the cultivation, storage, and transportation of pure cultures of bacteria.
Certifications: BSE Free