Triple Sugar Iron Agar, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C7110 C7111 C7112 C7113
CA89406-150EA 213.22 CAD
CA89406-150 CA89406-152 CA89406-154 CA89406-156
Triple Sugar Iron Agar, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Triple Sugar Iron (TSI) Agar is recommended for use in the differentiation of Enterobacteriaceae by their ability to ferment glucose, lactose, and sucrose, and produce hydrogen sulfide.
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Certifications: BSE Free