Tergitol 7 Agar, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C7050
C7051
C7052
C7053
CA89406-118EA
162.74
CAD
CA89406-118
CA89406-120
CA89406-122
CA89406-124
Tergitol 7 Agar, CRITERION™, Hardy Diagnostics
Media for Microbiology
Dehydrated Media for Microbiology
CRITERION™ Tergitol 7 Agar is used for enumerating and differentiating coliform bacteria
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
TTC Solution (0.05%), product 89407-742, can be added to this medium. Dehydratedculture media with engineered container.
Certifications: BSE Free