TCBS Agar, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C7040 C7041
CA89406-110EA 316.46 CAD
CA89406-110 CA89406-112
TCBS Agar, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ TCBS (Thiosulfate Citrate Bile Salts Sucrose) Agar is recommended for the selective isolation and cultivation of Vibrio spp
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
from clinical specimens. Dehydratedculture media with engineered container.
Certifications: BSE Free