TAT Broth Base, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C7030 C7031 C7032 C7033
CA89406-102EA 155.65 CAD
CA89406-102 CA89406-104 CA89406-106 CA89406-108
TAT Broth Base, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ TAT Broth Base is used for cultivating microorganisms from highly viscous or gelatinous materials
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Tween, product 89407-642, is required for this medium. Dehydratedculture media with engineered container.
Certifications: BSE Free