CRITERION™ Sucrose
Supplier: HARDY DIAGNOSTICS CA
C7020 C7021 C7022 C7023
CA89406-094EA 159.59 CAD
CA89406-094 CA89406-096 CA89406-098 CA89406-100
CRITERION™ Sucrose
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Sucrose is added to culture media for use in preparing microbiological culture media.
  • Wide mouth opening
  • Reduces dust and allows easy scooping
  • Convenient handgrip
  • 'No slip' finger holds
  • Stackable containers
  • High solubility; dissolves quickly
  • Custom and bulk products available
  • Unparalleled performance - tested for optimal growth
For fermentation tests it is usually added at a final concentration of 0.5 to 1.0%. Dehydratedculture media with engineered container.
These culture media is formulated to meet or exceed the highest standards.
Application: Breweries, wineries and distilleries, cannabis QA testing, clinical, cosmetics, environmental, food safety, compounding pharmacy, pharmaceutical manufacturing
Certifications: TSE/BSE free.
Delivery information: 500 g - wide mouth, easy grip bottle, comes with a free scoop, 2 and 10 kg bucket - with convenient screw lid.