Starch Agar, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C7010 C7011 C7012 C7013
CA89406-086EA 174.54 CAD
CA89406-086 CA89406-088 CA89406-090 CA89406-092
Starch Agar, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Starch Agar is used for cultivating microorganisms being tested for starch hydrolysis
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free