Standard Methods Agar, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C6980 C6981 C6982 C6983
CA89406-062EA 188.16 CAD
CA89406-062 CA89406-064 CA89406-066 CA89406-068
Standard Methods Agar, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Standard Methods Agar (Plate Count Agar) is recommended for use in determining the microbial content in dairy products, food, water and other material of sanitary importance
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free