Simmons Citrate Agar, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C6950 C6951 C6952 C6953
CA89406-046EA 189.05 CAD
CA89406-046 CA89406-048 CA89406-050 CA89406-052
Simmons Citrate Agar, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Simmons Citrate Agar is recommended for the differentiation of gram-negative enteric bacteria based on citrate utilization
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free