Salmonella Shigella Agar (SS), CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C6840 C6841
CA89406-006EA 479.44 CAD
CA89406-006 CA89406-008 CA89406-010 CA89406-012
Salmonella Shigella Agar (SS), CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Salmonella Shigella (SS) Agar is a highly selective agar used for the isolation of Salmonella and some Shigella
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free