Rose Bengal Agar Base, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C8101 C8102
CA89406-686EA 414.99 CAD
CA89406-686 CA89406-688
Rose Bengal Agar Base, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Mycobiotic Agar is recommended for use in the selective isolation and recovery of pathogenic fungi and dermatophytes from clinical specimens; especially hair, skin, and nails
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free