Selenite Cystine Broth, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C6920 C6921 C6922 C6923
CA89406-022EA 183.04 CAD
CA89406-022 CA89406-024 CA89406-026 CA89406-028
Selenite Cystine Broth, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Selenite Cystine Broth is for selective cultivation of Salmonella spp. from food, feces, dairy products and other sanitary materials
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free