Selective Strep Agar Base, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C6890 C6891 C6892 C6893
CA89406-014EA 188.38 CAD
CA89406-014 CA89406-016 CA89406-018 CA89406-020
Selective Strep Agar Base, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Selective Strep Agar Base with added sheep blood, this medium is recommended for the selective isolation and differentiation of pathogenic streptococci
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free