Sabdex Agar, Emmon's, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C6770 C6771 C6772 C6773
CA89405-966EA 177.69 CAD
CA89405-966 CA89405-968 CA89405-970 CA89405-972
Sabdex Agar, Emmon's, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Sabouraud Dextrose Agar (Sabdex), Emmons is recommended for the isolation, cultivation, and maintenance of non-pathogenic and pathogenic species of fungi and yeasts
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free