CRITERION™ Purple Broth Base
Supplier: HARDY DIAGNOSTICS CA
C6710 C6711
CA89405-944EA 263.47 CAD
CA89405-944 CA89405-946
CRITERION™ Purple Broth Base
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ purple broth base is primarily for use with added carbohydrate in differentiating pure cultures of bacteria, particularly of enteric organisms, based on fermentation reactions.
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
  • Unparalleled performance - tested for optimal growth
Dehydrated culture media with engineered container.
These culture media is formulated to meet or exceed the highest standards.
Application: Breweries, wineries and distilleries, cannabis QA testing, clinical, cosmetics, environmental, food safety, compounding pharmacy, pharmaceutical manufacturing.
Certifications: BSE free.
Delivery information: 500 g - wide mouth, easy grip bottle, comes with a free scoop, 2 and 10 kg bucket - with convenient screw lid.