Phenylalanine Agar, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C6600
C6601
C6602
C6603
CA89405-882EA
200.87
CAD
CA89405-882
CA89405-884
CA89405-886
CA89405-888
Phenylalanine Agar, CRITERION™, Hardy Diagnostics
Media for Microbiology
Dehydrated Media for Microbiology
CRITERION™ Phenylalanine Agar for the differentiation of gram negative bacilli based on phenylalnine deamination
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free