Phenol Red Broth Base, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C6590 C6591 C6592 C6593
CA89405-874EA 246.57 CAD
CA89405-874 CA89405-876 CA89405-878 CA89405-880
Phenol Red Broth Base, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Phenol Red Broth Base is intended to be used in the differentiation of microorganisms based on fermentation reactions
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free
Order Now
SPECIFICATIONS
Environmentally Preferable
Storage temperature –2-30°C