Modified Casman, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C7620 C7621 C7622 C7623
CA89406-444EA 192.79 CAD
CA89406-444 CA89406-446 CA89406-448 CA89406-450
Modified Casman, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Modified Casman for the cultivation of Haemophilus spp
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
and with added sheep blood can be used for the differentiation of Haemophilus spp. Dehydratedculture media with engineered container.
Certifications: BSE Free