MIO Medium, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C6330 C6331 C6332 C6333
CA89405-726EA 198.41 CAD
CA89405-726 CA89405-728 CA89405-730 CA89405-732
MIO Medium, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ MIO Medium is recommended for use in testing motility, indole production, and ornithine decarboxylase activity of enteric bacilli
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free
Order Now
SPECIFICATIONS
Environmentally Preferable
Storage temperature –2-30°C