McClung Toabe Agar Base, CRITERION™ Dehydrated Culture Media, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C8250 C8251
CA89406-738EA 299.63 CAD
CA89406-738 CA89406-740
McClung Toabe Agar Base, CRITERION™ Dehydrated Culture Media, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ McClung Toabe Agar Base is recommended for the isolation and cultivation of Clostridium perfringens in foods
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free