m FC Agar Base, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C6270
C6271
C6272
C6273
CA89405-684EA
219.39
CAD
CA89405-684
CA89405-686
CA89405-688
CA89405-690
m FC Agar Base, CRITERION™, Hardy Diagnostics
Media for Microbiology
Dehydrated Media for Microbiology
CRITERION™ m FC Agar Base is primarily for use with added Rosolic Acid for the cultivation and enumeration of fecal coliforms using membrane filtration technique at elevated temperatures
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free