Luria Agar Base, Miller, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C6310 C6311 C6312 C6313
CA89405-710EA 160.54 CAD
CA89405-710 CA89405-712 CA89405-714 CA89405-716
Luria Agar Base, Miller, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Luria Agar Base, Miller is used for maintaining and propagating Escherichia coli in molecular microbiology procedures with or without added glucose
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free