Lowenstein Jensen Medium Base, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C7380 C7381 C7382 C7383
CA89406-316EA 195.27 CAD
CA89406-316 CA89406-318 CA89406-320 CA89406-322
Lowenstein Jensen Medium Base, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Lowenstein Medium Base, when supplemented with egg and glycerol, is used for the cultivation, isolation, and differentiation of Mycobacteria spp. Glycerol, product #228210, and eggs are required for this medium. Dehydratedculture media with engineered container.
· Wide mouth opening - reduces dust formation by allowing the easy entry of a scoop
· Convenient handgrip - 'no slip' finger holds
· Stackable containers - for efficient storage
· Increased solubility - dissolves quickly
· Large selection, over 200 formulas available
· Custom and bulk products available - to your specifications
· Tested for optimal performance
· BSE Free (contact for specifics)
· Over 30 years of formulation experience
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Custom and bulk products available. BSE Free documentation available. Over 30 years of formulation experience
CRITERION™ Lowenstein Medium Base, when supplemented with egg and glycerol, is used for the cultivation, isolation, and differentiation of Mycobacteria spp
Glycerol, product #228210, and eggs are required for this medium. Dehydratedculture media with engineered container.
Certifications: BSE Free