Letheen Broth, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C6020
C6021
C6022
C6023
CA89405-580EA
171
CAD
CA89405-580
CA89405-582
CA89405-584
CA89405-586
Letheen Broth, CRITERION™, Hardy Diagnostics
Media for Microbiology
Dehydrated Media for Microbiology
CRITERION™ Letheen Broth is used to determine the antimicrobial activity of quaternary ammonium compounds
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free