Kligler Iron Agar, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C5910 C5911 C5912 C5913
CA89405-512EA 178.95 CAD
CA89405-512 CA89405-514 CA89405-516 CA89405-518
Kligler Iron Agar, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Kligler Iron Agar (KIA) is recommended for use in differentiating certain members of the Enterobacteriaceae by demonstrating hydrogen sulfide production and the fermentation of dextrose and lactose
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free