Inhibitory Mold Agar, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C5890 C5891 C5892 C5893
CA89405-504EA 187.83 CAD
CA89405-504 CA89405-506 CA89405-508 CA89405-510
Inhibitory Mold Agar, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Inhibitory Mold Agar is recommended for the selective isolation of fungi
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free