Group A Beta Strep Agar Base, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C5110 C5111 C5112 C5113
CA89405-120EA 378.11 CAD
CA89405-120 CA89405-122 CA89405-124 CA89405-126
Group A Beta Strep Agar Base, CRITERION™, Hardy Diagnostics
Media for Microbiology Prepared Media for Microbiology
CRITERION™ Group A Beta Strep Agar Base is an enriched media for the selective isolation of group A Streptococcus (Streptococcus pyogenes) from clinical specimens
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free