Gelatin, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C7920
C7921
C7922
C7923
CA89406-576EA
166.66
CAD
CA89406-576
CA89406-578
CA89406-580
CA89406-582
Gelatin, CRITERION™, Hardy Diagnostics
Media for Microbiology
Dehydrated Media for Microbiology
CRITERION™ Gelatin is primarily a high grade preparation of gelatin
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
It can be used as a solidification agent, incorporated into microbiological culture media as a nutrient, or to determine gelatinase activity (gelatin liquefaction) by microorganisms.1.0%. Dehydratedculture media with engineered container.
Certifications: BSE Free