GC Agar Base, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C5790 C5791 C5792 C5793
CA89405-456EA 194.68 CAD
CA89405-456 CA89405-458 CA89405-460 CA89405-462
GC Agar Base, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ GC Agar Base is used in the preparation of media for the cultivation and selective isolation of fastidious organisms such as Neisseria and Haemophilus species
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
VCAT Selective Supplement, product #SR104B and GC Growth Supplement, Vitox with Diluent, product #SR090A, are required for this medium. Dehydratedculture media with engineered container.
Certifications: BSE Free