Esculin Mannitol Agar, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C7590 C7591
CA89406-420EA 308.64 CAD
CA89406-420 CA89406-422 CA89406-424 CA89406-426
Esculin Mannitol Agar, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Esculin Mannitol Agar for the selective isolation and presumptive identification of Staphylococcus aureus and group D streptococci
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free