EMB Agar, Levine, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C5700 C5701 C5702 C5703
CA89405-428EA 196.69 CAD
CA89405-428 CA89405-430 CA89405-432 CA89405-434
EMB Agar, Levine, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ EMB Agar, Levine is recommended for use as a selective and differential medium for the isolation of gram-negative bacilli (including coliform organisms and enteric pathogens)
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free