CRITERION™ Dextrose (Glucose), Dehydrated Culture Media
Supplier: HARDY DIAGNOSTICS CA
C5580
C5581
C5582
C5583
CA89405-370EA
181.04
CAD
CA89405-370
CA89405-372
CA89405-374
CA89405-376
CRITERION™ Dextrose (Glucose), Dehydrated Culture Media
Media for Microbiology
Dehydrated Media for Microbiology
CRITERION™ Dextrose (Glucose) is chemically pure monosaccharide.
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
It is added to culture media as a source of energy for bacteria and for the determination of fermentation reactions. Dehydratedculture media with engineered container.
Certifications: BSE Free.