Corn Meal Agar, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C5490 C5491 C5492 C5493
CA89405-322EA 185.37 CAD
CA89405-322 CA89405-324 CA89405-326 CA89405-328
Corn Meal Agar, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Corn Meal Agar is recommended for use in the cultivation of fungi and for the inducement of chlamydospore formation by Candida albicans
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
CRITERION™ Dehydratedculture media with engineered container.
Certifications: BSE Free