Columbia CNA Agar Base, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C5470 C5471 C5472 C5473
CA89405-306EA 192.29 CAD
CA89405-306 CA89405-308 CA89405-310 CA89405-312
Columbia CNA Agar Base, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Columbia CNA Agar Base is intended to be used as a selective basal media to which blood is added for the selective isolation and differentiation of gram-positive cocci.
Dehydratedculture media with engineered container.
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Custom and bulk products available. BSE Free documentation available. Over 30 years of formulation experience
CRITERION™ Columbia CNA Agar Base is intended to be used as a selective basal media to which blood is added for the selective isolation and differentiation of gram-positive cocci.
Certifications: BSE Free