Cetrimide Selective Agar, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C5370 C5371 C5372 C5373
CA89405-274EA 193.98 CAD
CA89405-274 CA89405-276 CA89405-278 CA89405-280
Cetrimide Selective Agar, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Cetrimide Selective Agar is primarily for use in the isolation and detection of Pseudomonas aeroginosa
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free