Caffeic Acid Agar, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C7780 C7781
CA89406-520EA 250.76 CAD
CA89406-520 CA89406-522
Caffeic Acid Agar, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Caffeic Acid Agar is recommended for the selective isolation and differentiation of Cryptococcus neoformans
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free