CRITERION™ Buffered Peptone Water
Supplier: HARDY DIAGNOSTICS CA
C5320 C5321 C5322 C5323 C5324
CA89405-248EA 164.45 CAD
CA89405-248 CA89405-250 CA89405-252 CA89405-254 CA89405-256
CRITERION™ Buffered Peptone Water
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ buffered peptone water is intended to aid in the recovery of injured Salmonella species from foods and other samples prior to selective enrichment and isolation. Dehydratedculture media with engineered container.
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Custom and bulk products available. BSE Free documentation available. Over 30 years of formulation experience
Certifications: BSE Free.