Brilliant Green Agar, Modified, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C8690
C8691
C8692
C8693
CA89406-752EA
196.6
CAD
CA89406-752
CA89406-754
CA89406-756
CA89406-758
Brilliant Green Agar, Modified, CRITERION™, Hardy Diagnostics
Media for Microbiology
Dehydrated Media for Microbiology
CRITERION™ Brilliant Green Agar, Modified, is for the selective isolation of Salmonella, except for Salmonella typhi
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free