Brilliant Green Agar, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C5240 C5241 C5242 C5243
CA89405-208EA 202.18 CAD
CA89405-208 CA89405-210 CA89405-212 CA89405-214
Brilliant Green Agar, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Brilliant Green Agar is recommended for the selective enrichment of Salmonella spp
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
other than S. typhi and S. paratyphi from clinical and non-clinical specimens. Dehydratedculture media with engineered container.
Certifications: BSE Free
Order Now
SPECIFICATIONS
Environmentally Preferable
Storage temperature –2-30°C