Brain Heart Infusion Agar, CRITERION™ Dehydrated Culture Media, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C5120 C5121 C5122 C5123
CA89405-128EA 226.22 CAD
CA89405-128 CA89405-130 CA89405-132 CA89405-134
Brain Heart Infusion Agar, CRITERION™ Dehydrated Culture Media, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Brain Heart Infusion Agar is a general purpose nutrient medium recommended for the cultivation and isolation of a variety of fastidious and nonfastidious microorganisms, including bacteria, yeasts, and mold
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free