CRITERION™ Bismuth Sulfite Agar
Supplier: HARDY DIAGNOSTICS CA
C5210 C5211 C5212 C5213
CA89405-192EA 259.27 CAD
CA89405-192 CA89405-194 CA89405-196 CA89405-198
CRITERION™ Bismuth Sulfite Agar
Media for Microbiology Dehydrated Media for Microbiology
Bismuth sulfite agar for the selective isolation of Salmonella spp from clinical and food specimens. Dehydratedculture media with engineered container.
  • Wide mouth opening - reduces dust formation by allowing the easy entry of a scoop
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers - for efficient storage
  • Increased solubility - dissolves quickly
  • Large selection, over 200 formulas available
Certifications: BSE Free.