Bile Salts, #3, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C5200 C5201
CA89405-184EA 687.3 CAD
CA89405-184 CA89405-186
Bile Salts, #3, CRITERION™, Hardy Diagnostics
Media for Microbiology Additives and Supplements for Microbiology
CRITERION™ Bile Salts #3 is specifically formulated for use in MacConkey Agar, SS Agar, and Violet Red Bile Agar
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free