Bile Esculin Azide Agar, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C5170
C5171
C5172
C5173
CA89405-160EA
286.23
CAD
CA89405-160
CA89405-162
CA89405-164
CA89405-166
Bile Esculin Azide Agar, CRITERION™, Hardy Diagnostics
Media for Microbiology
Dehydrated Media for Microbiology
CRITERION™ Bile Esculin Azide Dehydrated Culture Media are recommended for the isolation and differentiation of group D from non-group D streptococci
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
CRITERION™ Dehydratedculture media with engineered container.
Certifications: BSE Free