Baird Parker Agar Base, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C5080 C5081 C5082 C5083
CA89405-104EA 218.55 CAD
CA89405-104 CA89405-106 CA89405-108 CA89405-110
Baird Parker Agar Base, CRITERION™, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ Baird-Parker Agar Base is used with Egg Yolk Tellurite Enrichment as a selective medium for the detection and enumeration of coagulase-positive staphylococci from food samples
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Egg Yolk Tellurite Emulsion, product 89407-702, is required for this meduim. Dehydratedculture media with engineered container.
Certifications: BSE Free