CulGenex™ Yeast Peptone Dextrose (YPD), Dehydrated Culture Media, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
C1159 CG91
CA89405-044PK 2944.27 CAD
CA89405-044 CA89407-088
CulGenex™ Yeast Peptone Dextrose (YPD), Dehydrated Culture Media, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CulGenex™ Yeast Peptone Dextrose (YPD) Powders are recommended for the rapid growth of yeasts, particularly Saccharomyces cerevisiae, used for molecular research.