Cutters, Tubing, Excelta
Supplier: Excelta
2922-14-X 2922-14 2922-18 2922-38
89411-712EA 24.46 CAD
89411-712 89411-842 89411-844 75880-684 76250-660
Cutters, Tubing, Excelta
Cutters
Gives a nice clean cut to a variety of soft tubing materials.
  • Medical 'clean-cut' tubing cutter
  • Replaceable blades
  • Ergonomic grips and dual leaf springs
The 2922 Series and the 51-T medical tubing cutter give a nice clean cut to a variety of soft tubing materials. The blades on the 2922 series are replaceable and the grips are soft cushioned foam and have a surface resistivity of 10¹⁰ ohms/sq.