Mantles, Fabric/Cloth, Ace Glass Incorporated
Supplier: Ace Glass
12036-17 12036-19 12036-21 12036-23 12036-24
89075-880EA 1376.54 CAD
89075-880 89075-882 89075-884 89075-886 89075-888
Mantles, Fabric/Cloth, Ace Glass Incorporated
Heaters Heating Mantles
Heating mantles are designed for use with reaction flasks
Upper temperature limit is 450°C (842°F). A 122cm (4') cord and locking connector are included. Units must be operated through a temperature controller.
Certifications: CSA approved.