Trubore® Bearing , Economy, Ace Glass Incorporated
Supplier: Ace Glass
9529-04 9529-07 9529-10
89061-842EA 115.49 CAD
89061-842 89075-066 89075-068
Trubore® Bearing , Economy, Ace Glass Incorporated
Stirrers Stirring Tools
Trubore® bearing, heavy duty, standard taper joint, self-aligning
Unique design is forgiving of slight misalignment of stir motor and shaft. Recommended for use with ACE 6 mm precision ground stir shaft.