High Temperature Dry Block Heater, BT5D, Grant Instruments
Supplier: GRANT USA INC.
BT5D-16L BT5D-26L
71003-686EA 9306.2 CAD
71003-686 89498-130
High Temperature Dry Block Heater, BT5D, Grant Instruments
Block Heaters Digital Block Heaters
High temperature digital block heaters with single fixed aluminium block
  • Operating temperature range is ambient +10 to 400°C
  • Temperature stability at 300°C is ±0,5°C and temperature uniformity is ±1% (maximum temperature difference between tubes: 2°C at 200°C, 3°C at 300°C, 4°C at 400°C)
  • Timer for operation from 1 to 9999 min., or continuous operation
  • Ideal for use at high temperatures - avoids the use of oil and laboratory remains steam-free
  • Adjustable overtemperature cut-out
  • Choice of two models with different block capacities
The BT5D is ideal for any application requiring heating in a dry block up to 400ºC.
Units feature microprocessor control, digital temperature settings and display, high and low alarms, and a controlled heating rate. Heaters have LED with 0,1°C setting resolution and 1°C display resolution.
Certifications: CE Approved.
Ordering information: Choice of two models: BT5D-16 for 38 x Ø 16 mm tubes and BT5D-26 for 22 x Ø 26 mm tubes. Other sizes also available; please contact your Avantor representative for more information.