MRI Core DX Carts, Polyethylene, TrippNT
Supplier: TrippNT
51728SMOKE 51728BLUE 51728
10754-750EA 3300.95 CAD
10754-750 10754-606 89408-474
MRI Core DX Carts, Polyethylene, TrippNT
Trolleys
Non-ferrous MRI Core DX Lab Cart offers a MR Conditional organizing cart expands storage needs with drawers, shelves and bulk storage offers flexible storage as your needs change.
  • Non-ferrous MRI organizational chart
  • 11 flexible storage features
  • 3 drawers
  • 6 side shelves
  • Bulk storage area
  • Tool holder
  • Chemically resistant polyethylene construction
  • Top has two built in handles making it smooth to maneuver
  • 3 inch casters
  • Fully assembled, ready to use out of the box
  • American made by TrippNT
  • Total Replacement Lifetime Guarantee
Made from food grade high density polyethylene and ABS. Completely non-magnetic easy rolling cart with 3 large drawers, 6 side shelves and a bulk storage item.